4-Hydroxybenzoylacetonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(Cyanoacetyl)phenol, 3-(4-Hydroxyphenyl)-3-oxopropanenitrile
OR7015-250MG
OR7015-1G
APOSOR7015-250MGEA
7940
CZK
APOSOR7015-250MG
APOSOR7015-1G
4-Hydroxybenzoylacetonitrile
4-Hydroxybenzoylacetonitrile
Vzorec:
C₉H₇NO₂ MW: 161,157 g/mol Bod tání: 189…192 °C Storage Temperature: Ambient |
MDL Number:
MFCD07786224 CAS číslo: 70591-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...