Tisk…

6-Bromo-4-chloro-1H-benzimidazole

Dodavatel: Apollo Scientific
Varování

OR70112-1G OR70112-250MG OR70112-5G
APOSOR70112-1GEA 1820 CZK
APOSOR70112-1G APOSOR70112-250MG APOSOR70112-5G
6-Bromo-4-chloro-1H-benzimidazole
6-Bromo-4-chloro-1H-benzimidazole
Vzorec: C₇H₄BrClN₂
MW: 231,477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22378839
CAS číslo: 1360932-45-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR