2-Cyclopropylphenol 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(2-Hydroxyphenyl)cyclopropane
OR70019-1G
OR70019-250MG
APOSOR70019-1GEA
14150
CZK
APOSOR70019-1G
APOSOR70019-250MG
2-Cyclopropylphenol 98%
2-Cyclopropylphenol
Vzorec:
C₉H₁₀O MW: 134,175 g/mol Bod varu: 79…80 °C (2 mm Hg) Bod vzplanutí: >70 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06802666 CAS číslo: 10292-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...