Tisk…

5-Ethylcyclohexane-1,3-dione 98%

Dodavatel: Apollo Scientific
Varování

OR6970-1G OR6970-250MG OR6970-500MG
APOSOR6970-1GEA 7870 CZK
APOSOR6970-1G APOSOR6970-250MG APOSOR6970-500MG
5-Ethylcyclohexane-1,3-dione 98%
5-Ethylcyclohexane-1,3-dione
Vzorec: C₈H₁₂O₂
MW: 140,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129457
CAS číslo: 862168-16-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR