1-(3-Chloroprop-1-yl)piperazine dihydrochloride hemihydrate 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR6879-100MG
OR6879-500MG
APOSOR6879-100MGEA
1240
CZK
APOSOR6879-100MG
APOSOR6879-500MG
1-(3-Chloroprop-1-yl)piperazine dihydrochloride hemihydrate 95%
1-(3-Chloroprop-1-yl)piperazine dihydrochloride hemihydrate
Vzorec:
C₇H₁₅ClN₂·2ClH·H₂O Bod tání: ∼ 215 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150090 CAS číslo: 34782-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...