Tisk…

1-(1-Phenylethyl)piperazine 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: [1-(Piperazin-1-yl)ethyl]benzene

OR6850-1G
APOSOR6850-1GEA 1490 CZK
APOSOR6850-1G
1-(1-Phenylethyl)piperazine 97%
1-(1-Phenylethyl)piperazine
Vzorec: C₁₂H₁₈N₂
MW: 190,285 g/mol
Bod varu: 91…94 °C (0,1 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00040738
CAS číslo: 69628-75-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR