Tisk…

N-Methyl-1-[4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]methylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-Methyl-3-{4-[(methylamino)methyl]phenyl}-1,2,4-oxadiazole, N-Methyl-4-(5-methyl-1,2,4-oxadiazol-3-yl)benzylamine

OR6728-100MG OR6728-1G OR6728-250MG
APOSOR6728-100MGEA 1660 CZK
APOSOR6728-100MG APOSOR6728-1G APOSOR6728-250MG
N-Methyl-1-[4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]methylamine 97%
N-Methyl-1-[4-(5-methyl-1,2,4-oxadiazol-3-yl)phenyl]methylamine
Vzorec: C₁₁H₁₃N₃O
Bod tání: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD07772819
CAS číslo: 857283-77-3
UN: 2923
ADR: 8+6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR