5-Methyl-3-phenylisoxazole-4-sulphonyl chloride 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6722-1G
APOSOR6722-1GEA
0
CZK
APOSOR6722-1G
5-Methyl-3-phenylisoxazole-4-sulphonyl chloride 97%
5-Methyl-3-phenylisoxazole-4-sulphonyl chloride
Vzorec:
C₁₀H₈ClNO₃S MW: 257,693 g/mol Bod tání: 61…62 °C Storage Temperature: Ambient |
MDL Number:
MFCD06658966 CAS číslo: 857283-56-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...