Tisk…

1-(4-Isocyanatophenyl)-1H-pyrrole 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(1H-Pyrrol-1-yl)phenyl isocyanate

OR6721-1G OR6721-250MG
APOSOR6721-1GEA 28070 CZK
APOSOR6721-1G APOSOR6721-250MG
1-(4-Isocyanatophenyl)-1H-pyrrole 97%
1-(4-Isocyanatophenyl)-1H-pyrrole

When melted, may contain small amount of sediment

Vzorec: C₁₁H₈N₂O
MW: 184,194 g/mol
Bod tání: 65…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07772803
CAS číslo: 857283-60-4
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR