Tisk…

3,5-Dibromophenylacetic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 2-(3,5-Dibromophenyl)acetic acid

OR6636-10G OR6636-25G
APOSOR6636-10GEA 0 CZK
APOSOR6636-10G APOSOR6636-25G
3,5-Dibromophenylacetic acid 98%
3,5-Dibromophenylacetic acid
Vzorec: C₈H₆Br₂O₂
MW: 293,94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787513
CAS číslo: 188347-49-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR