Tisk…

(1E)-1-Chloroprop-1-ene 96%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: (1E)-Prop-1-en-1-yl chloride, trans-1-Chloroprop-1-ene

OR6595-25G OR6595-5G
APOSOR6595-25GEA 27350 CZK
APOSOR6595-25G APOSOR6595-5G
(1E)-1-Chloroprop-1-ene 96%
(1E)-1-Chloroprop-1-ene
Vzorec: C₃H₅Cl
MW: 76,525 g/mol
Bod varu: 38…39 °C
Hustota: 0,94 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039358
CAS číslo: 16136-85-9
UN: 1992
ADR: 3+6.1,I

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR