Tisk…

3-Thien-3-ylacrylic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Thien-3-yl)prop-2-enoic acid, 3-(2-Carboxyethenyl)thiophene, (E)-3-(Thiophen-3-yl)acrylic acid, 3-(Thien-3-yl)acrylic acid

OR6590-10G OR6590-5G
APOSOR6590-10GEA 5000 CZK
APOSOR6590-10G APOSOR6590-5G
3-Thien-3-ylacrylic acid 97%
3-Thien-3-ylacrylic acid
Vzorec: C₇H₆O₂S
MW: 154,19 g/mol
Bod tání: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00014535
CAS číslo: 1195-52-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR