Tisk…

3-(Thien-2-yl)acrylic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(2-Carboxyethenyl)thiophene, 3-(Thiophen-2-yl)acrylic acid, 3-(Thien-2-yl)prop-2-enoic acid

OR6589-25G OR6589-5G
APOSOR6589-25GEA 2780 CZK
APOSOR6589-25G APOSOR6589-5G
3-(Thien-2-yl)acrylic acid 98%
3-(Thien-2-yl)acrylic acid
Vzorec: C₇H₆O₂S
MW: 154,186 g/mol
Bod tání: 145…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005455
CAS číslo: 1124-65-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR