2-Bromooctadecanoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR6493-1G
OR6493-5G
APOSOR6493-1GEA
4140
CZK
APOSOR6493-1G
APOSOR6493-5G
2-Bromooctadecanoic acid
2-Bromooctadecanoic acid
Radiopharmaceutical precursor
Vzorec:
C₁₈H₃₅BrO₂ Bod tání: 58…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00021694 CAS číslo: 142-94-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...