Tisk…

6-Bromo-2-chloroacridine-9-carbonitrile

Dodavatel: Apollo Scientific
Varování

OR64742-1G OR64742-250MG OR64742-5G
APOSOR64742-1GEA 3110 CZK
APOSOR64742-1G APOSOR64742-250MG APOSOR64742-5G
6-Bromo-2-chloroacridine-9-carbonitrile
6-Bromo-2-chloroacridine-9-carbonitrile
Vzorec: C₁₄H₆BrClN₂
MW: 317,568 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18823120
CAS číslo: 958452-43-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR