Tisk…

2-Bromoacridine-9-carbonitrile

Dodavatel: Apollo Scientific
Varování

OR64741-1G OR64741-250MG OR64741-5G
APOSOR64741-1GEA 3060 CZK
APOSOR64741-1G APOSOR64741-250MG APOSOR64741-5G
2-Bromoacridine-9-carbonitrile
2-Bromoacridine-9-carbonitrile
Vzorec: C₁₄H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD32066850
CAS číslo: 958452-37-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR