Tisk…

3-Bromobenzylguanidinium sulphate

Dodavatel: Apollo Scientific
Varování

OR6468-100MG OR6468-200MG
APOSOR6468-100MGEA 7540 CZK
APOSOR6468-100MG APOSOR6468-200MG
3-Bromobenzylguanidinium sulphate
3-Bromobenzylguanidinium sulphate

Radiopharmaceutical precursor

Vzorec: C₈H₁₀BrN₃·H₂SO₄
MW: 326.168 g/mol
Bod tání: 174…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00040820
CAS číslo: 90151-50-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR