6-Chloro-3-nitropyridine-2-carbonitrile 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-3-nitropicolinonitrile, 6-Chloro-2-cyano-3-nitropyridine
OR6409-1G
OR6409-25G
OR6409-5G
APOSOR6409-1GEA
622
CZK
APOSOR6409-1G
APOSOR6409-25G
APOSOR6409-5G
6-Chloro-3-nitropyridine-2-carbonitrile 98%
6-Chloro-3-nitropyridine-2-carbonitrile
Vzorec:
C₆H₂ClN₃O₂ MW: 183,552 g/mol Bod tání: 109…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD06657641 CAS číslo: 93683-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...