3-(Azetidin-3-yl)propan-1-ol hydrochloride
Dodavatel: Apollo Scientific
Synonyms:
hydrochloride, 3-(Azetidin-3-yl)propan-1-ol
OR63047-1G
OR63047-250MG
APOSOR63047-1GEA
19840
CZK
APOSOR63047-1G
APOSOR63047-250MG
3-(Azetidin-3-yl)propan-1-ol hydrochloride
3-(Azetidin-3-yl)propan-1-ol hydrochloride
Vzorec:
C₆H₁₃NO·ClH MW: 151,634 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD18206286 CAS číslo: 1379237-95-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...