4-Bromo-3-iodobenzaldehyde
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR63029-250MG
OR63029-1G
OR63029-5G
APOSOR63029-250MGEA
1660
CZK
APOSOR63029-250MG
APOSOR63029-1G
APOSOR63029-5G
4-Bromo-3-iodobenzaldehyde
4-Bromo-3-iodobenzaldehyde
Vzorec:
C₇H₄BrIO MW: 310.915 g/mol Bod tání: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD18391620 CAS číslo: 873387-81-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...