3-(4-Iodopyrazol-1-yl)propanoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR62032-1G
OR62032-250MG
OR62032-5G
APOSOR62032-1GEA
4800
CZK
APOSOR62032-1G
APOSOR62032-250MG
APOSOR62032-5G
3-(4-Iodopyrazol-1-yl)propanoic acid
3-(4-Iodopyrazol-1-yl)propanoic acid
Vzorec:
C₆H₇IN₂O₂ MW: 266,036 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD15146427 CAS číslo: 6715-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...