Tisk…

2-(4-Iodopyrazol-1-yl)acetonitrile

Dodavatel: Apollo Scientific
Nebezpečí

OR62031-1G OR62031-250MG OR62031-5G
APOSOR62031-1GEA 2110 CZK
APOSOR62031-1G APOSOR62031-250MG APOSOR62031-5G
2-(4-Iodopyrazol-1-yl)acetonitrile
2-(4-Iodopyrazol-1-yl)acetonitrile
Vzorec: C₅H₄IN₃
MW: 233,01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06014071
CAS číslo: 955965-81-8
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR