Tisk…

Ethyl 2-(4-iodopyrazol-1-yl)acetate

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl 2-(4-iodo-1H-pyrazol-1-yl)acetate

OR62029-1G OR62029-250MG OR62029-5G
APOSOR62029-1GEA 4900 CZK
APOSOR62029-1G APOSOR62029-250MG APOSOR62029-5G
Ethyl 2-(4-iodopyrazol-1-yl)acetate
Ethyl 2-(4-iodopyrazol-1-yl)acetate
Vzorec: C₇H₉IN₂O₂
MW: 280,063 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11054036
CAS číslo: 82231-59-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR