6-Chloro-5-methoxynicotinic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-5-methoxypyridine-3-carboxylic acid, 3-Carboxy-6-chloro-5-methoxypyridine
OR62002-100MG
OR62002-1G
OR62002-500MG
OR62002-250MG
APOSOR62002-100MGEA
0
CZK
APOSOR62002-100MG
APOSOR62002-1G
APOSOR62002-500MG
APOSOR62002-250MG
6-Chloro-5-methoxynicotinic acid 95%
6-Chloro-5-methoxynicotinic acid
Vzorec:
C₇H₆ClNO₃ MW: 187,58 g/mol Bod tání: 188…189 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD12828186 CAS číslo: 915107-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...