Tisk…

2,5-Dimethylthiophene-3-carboxaldehyde 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2,5-Dimethyl-3-formylthiophene

OR6179-5G OR6179-25G
APOSOR6179-5GEA 2440 CZK
APOSOR6179-5G APOSOR6179-25G
2,5-Dimethylthiophene-3-carboxaldehyde 95+%
2,5-Dimethylthiophene-3-carboxaldehyde
Vzorec: C₇H₈OS
Storage Temperature: Ambient
MDL Number: MFCD03426895
CAS číslo: 26421-44-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR