2-Bromo-4-(methylamino)-3-nitropyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-N-methyl-3-nitropyridin-4-amine
OR61612-1G
OR61612-250MG
APOSOR61612-1GEA
26450
CZK
APOSOR61612-1G
APOSOR61612-250MG
2-Bromo-4-(methylamino)-3-nitropyridine
2-Bromo-4-(methylamino)-3-nitropyridine
Vzorec:
C₆H₆BrN₃O₂ MW: 232.035 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22415286 CAS číslo: 1396554-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...