Tisk…

7-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine 95+%

Dodavatel: Apollo Scientific

Synonyms: 4-Chloro-2-methyl-7-azabenzimidazole

OR61522-100MG OR61522-250MG
APOSOR61522-100MGEA 5390 CZK
APOSOR61522-100MG APOSOR61522-250MG
7-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine 95+%
7-Chloro-2-methyl-3H-imidazo[4,5-b]pyridine
Vzorec: C₇H₆ClN₃
MW: 167,596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190300
CAS číslo: 1086423-62-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR