Tisk…

4-Methyl-3-oxopentanenitrile 97+%

Dodavatel: Apollo Scientific
Varování

OR61296-5G OR61296-25G
APOSOR61296-5GEA 5390 CZK
APOSOR61296-5G APOSOR61296-25G
4-Methyl-3-oxopentanenitrile 97+%
4-Methyl-3-oxopentanenitrile
Vzorec: C₆H₉NO
Storage Temperature: Ambient
MDL Number: MFCD05664144
CAS číslo: 29509-06-6
UN: 3276
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR