2-(2-Chlorophenyl)-2-methylpropanoic acid
Dodavatel: Apollo Scientific
Synonyms:
2-(2-Chlorophenyl)-2-methylpropionic acid
OR61217-1G
OR61217-250MG
APOSOR61217-1GEA
39350
CZK
APOSOR61217-1G
APOSOR61217-250MG
2-(2-Chlorophenyl)-2-methylpropanoic acid
2-(2-Chlorophenyl)-2-methylpropanoic acid
Vzorec:
C₁₀H₁₁ClO₂ MW: 198,646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11036920 CAS číslo: 69849-06-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...