Tisk…

4-(Methylsulphonyl)-2-nitrophenol 95+%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Hydroxy-5-(methylsulphonyl)nitrobenzene, 4-Hydroxy-3-nitrophenyl methyl sulphone

OR61203-1G OR61203-5G OR61203-25G
APOSOR61203-1GEA 662 CZK
APOSOR61203-1G APOSOR61203-5G APOSOR61203-25G
4-(Methylsulphonyl)-2-nitrophenol 95+%
4-(Methylsulphonyl)-2-nitrophenol
Vzorec: C₇H₇NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00085946
CAS číslo: 97-10-9
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR