3-Bromo-2-nitrobenzonitrile 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR61169-1G
OR61169-5G
OR61169-500MG
APOSOR61169-1GEA
2900
CZK
APOSOR61169-1G
APOSOR61169-5G
APOSOR61169-500MG
3-Bromo-2-nitrobenzonitrile 95%
3-Bromo-2-nitrobenzonitrile
Vzorec:
C₇H₃BrN₂O₂ MW: 227,015 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18392276 CAS číslo: 1261777-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...