Tisk…

2-Amino-5-(2,6-dimethylmorpholin-4-yl)-1,3,4-thiadiazole

Dodavatel: Apollo Scientific

Synonyms: 4-(5-Amino-1,3,4-thiadiazol-2-yl)-2,6-dimethylmorpholine, 5-(2,6-Dimethylmorpholin-4-yl)-1,3,4-thiadiazol-2-amine

OR61159-1G OR61159-250MG OR61159-5G
APOSOR61159-1GEA 8270 CZK
APOSOR61159-1G APOSOR61159-250MG APOSOR61159-5G
2-Amino-5-(2,6-dimethylmorpholin-4-yl)-1,3,4-thiadiazole
2-Amino-5-(2,6-dimethylmorpholin-4-yl)-1,3,4-thiadiazole
Vzorec: C₈H₁₄N₄OS
MW: 214,288 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06243036
CAS číslo: 923682-98-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR