Tisk…

3-Formyl-4-hydroxybenzonitrile

Dodavatel: Apollo Scientific

Synonyms: 5-Cyano-2-hydroxybenzaldehyde, 4-Cyano-2-hydroxyphenol

OR61136-1G OR61136-250MG
APOSOR61136-1GEA 4800 CZK
APOSOR61136-1G APOSOR61136-250MG
3-Formyl-4-hydroxybenzonitrile
3-Formyl-4-hydroxybenzonitrile
Vzorec: C₈H₅NO₂
MW: 147,131 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09839541
CAS číslo: 74901-29-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR