Tisk…

(3S)-3-Ethylmorpholine 97%

Dodavatel: Apollo Scientific

Synonyms: (S)-3-Ethylmorpholine

OR61132-250MG OR61132-1G OR61132-5G
APOSOR61132-250MGEA 4730 CZK
APOSOR61132-250MG APOSOR61132-1G APOSOR61132-5G
(3S)-3-Ethylmorpholine 97%
(3S)-3-Ethylmorpholine
Vzorec: C₆H₁₃NO
MW: 115.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11519097
CAS číslo: 748117-01-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR