2,2'-(Oxydimethanediyl)bis(4-methylquinazoline)
Dodavatel: Apollo Scientific
|
Varování
|
OR61080-1G
OR61080-250MG
APOSOR61080-1GEA
7870
CZK
APOSOR61080-1G
APOSOR61080-250MG
2,2'-(Oxydimethanediyl)bis(4-methylquinazoline)
2,2'-(Oxydimethanediyl)bis(4-methylquinazoline)
Vzorec:
C₂₀H₁₈N₄O MW: 330,383 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31726313 CAS číslo: 2387695-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...