Tisk…

3-Iodo-2-methylphenacyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(3-Iodo-2-methylpheny)-2-bromoethan-1-one

OR61076-1G OR61076-250MG OR61076-5G
APOSOR61076-1GEA 705 CZK
APOSOR61076-1G APOSOR61076-250MG APOSOR61076-5G
3-Iodo-2-methylphenacyl bromide
3-Iodo-2-methylphenacyl bromide
Vzorec: C₉H₈BrIO
MW: 338,968 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31726337
CAS číslo: 2408223-87-8
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR