Tisk…

Methyl 3-bromo-1H-indazole-7-carboxylate

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Bromo-7-(methoxycarbonyl)-1H-indazole

OR61031-1G OR61031-100MG
APOSOR61031-1GEA 2150 CZK
APOSOR61031-1G APOSOR61031-100MG
Methyl 3-bromo-1H-indazole-7-carboxylate
Methyl 3-bromo-1H-indazole-7-carboxylate
Vzorec: C₉H₇BrN₂O₂
MW: 255,068 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16556320
CAS číslo: 1257535-37-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR