6-Chloro-5-methylnicotinic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloro-5-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-3-methylpyridine
OR61022-1G
OR61022-500MG
APOSOR61022-1GEA
8930
CZK
APOSOR61022-1G
APOSOR61022-500MG
6-Chloro-5-methylnicotinic acid
6-Chloro-5-methylnicotinic acid
Vzorec:
C₇H₆ClNO₂ MW: 171,581 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD17214192 CAS číslo: 66909-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...