Tisk…

1-Benzyl-3-({[(tert-butyl)dimethylsilyl]oxy}methyl)piperidin-4-one 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Benzyl-3-(((tert-butyldimethylsilyl)oxy)methyl)piperidin-4-one, 1-Benzyl-3-{[(tert-butyldimethylsilyl)oxy]methyl}-4-oxopiperidine,[(1-Benzyl-4-oxopiperidin-3-yl)methoxy]tert-butyldimethylsilane

OR6091-1G OR6091-5G
APOSOR6091-1GEA 5660 CZK
APOSOR6091-1G APOSOR6091-5G
1-Benzyl-3-({[(tert-butyl)dimethylsilyl]oxy}methyl)piperidin-4-one 95%
1-Benzyl-3-({[(tert-butyl)dimethylsilyl]oxy}methyl)piperidin-4-one
Vzorec: C₁₉H₃₁NO₂Si
MW: 333,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09065031
CAS číslo: 325486-37-1
UN: 2922
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR