Tisk…

5-Bromo-2-[(methylamino)methyl]pyridine 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-(5-Bromopyridin-2-yl)-N-methylmethylamine

OR6090-1G OR6090-250MG
APOSOR6090-1GEA 10350 CZK
APOSOR6090-1G APOSOR6090-250MG
5-Bromo-2-[(methylamino)methyl]pyridine 95%
5-Bromo-2-[(methylamino)methyl]pyridine
Vzorec: C₇H₉BrN₂
MW: 201,064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09065029
CAS číslo: 915707-70-9
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR