Methyl 4-[(chlorosulphonyl)methyl]benzoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 4-((chlorosulphonyl)methyl)benzoate, [4-(Methoxycarbonyl)phenyl]methanesulphonyl chloride, Methyl 4-(chlorosulphonylmethyl)benzoate
OR6079-1G
OR6079-250MG
APOSOR6079-1GEA
3970
CZK
APOSOR6079-1G
APOSOR6079-250MG
Methyl 4-[(chlorosulphonyl)methyl]benzoate
Methyl 4-[(chlorosulphonyl)methyl]benzoate
Vzorec:
C₉H₉ClO₄S MW: 248.683 g/mol Bod tání: 115.5…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD07778383 CAS číslo: 130047-14-2 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...