Ethyl 2-benzoyl-3-(dimethylamino)acrylate, tech
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
Ethyl 2-[(dimethylamino)methylene]-3-oxo-3-phenylpropanoate, Ethyl 2-benzoyl-3-(dimethylamino)prop-2-enoate
OR60237-1G
OR60237-25G
OR60237-5G
APOSOR60237-1GEA
2480
CZK
APOSOR60237-1G
APOSOR60237-25G
APOSOR60237-5G
Ethyl 2-benzoyl-3-(dimethylamino)acrylate, tech
Ethyl 2-benzoyl-3-(dimethylamino)acrylate
Vzorec:
C₁₄H₁₇NO₃ MW: 247,29 g/mol Bod varu: 150…155 °C (0,5 mm Hg) Bod tání: 62…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00120477 CAS číslo: 66129-60-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...