5-Bromo-2-hydrazino-4-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(5-Bromo-4-methylpyridin-2-yl)hydrazine, (5-Bromo-4-methylpyridin-2-yl)hydrazine, 5-Bromo-2-hydrazino-4-picoline
OR60233-5G
OR60233-1G
APOSOR60233-5GEA
15540
CZK
APOSOR60233-5G
APOSOR60233-1G
5-Bromo-2-hydrazino-4-methylpyridine
5-Bromo-2-hydrazino-4-methylpyridine
Vzorec:
C₆H₈BrN₃ MW: 202,052 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09999921 CAS číslo: 913839-67-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...