Tisk…

2-Hydrazino-4-methylpyridine

Dodavatel: Apollo Scientific
Varování

OR60231-1G OR60231-5G
APOSOR60231-1GEA 3110 CZK
APOSOR60231-1G APOSOR60231-5G
2-Hydrazino-4-methylpyridine
2-Hydrazino-4-methylpyridine
Vzorec: C₆H₉N₃
Storage Temperature: Refrigerator
MDL Number: MFCD09863246
CAS číslo: 4931-00-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR