3-Bromo-2-hydrazino-5-methylpyridine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Bromo-5-methylpyridin-2-yl)hydrazine, 5-Bromo-6-hydrazino-3-picoline
OR60227-1G
OR60227-250MG
APOSOR60227-1GEA
910
CZK
APOSOR60227-1G
APOSOR60227-250MG
3-Bromo-2-hydrazino-5-methylpyridine
3-Bromo-2-hydrazino-5-methylpyridine
Vzorec:
C₆H₈BrN₃ MW: 202,052 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD10000011 CAS číslo: 1289007-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...