Tisk…

2-Hydrazino-3-methylpyrazine

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Hydrazino-3-methyl-1,4-diazine, 2-Hydrazineyl-3-methylpyrazine, (3-Methylpyrazin-2-yl)hydrazine

OR60215-1G OR60215-250MG
APOSOR60215-1GEA 10250 CZK
APOSOR60215-1G APOSOR60215-250MG
2-Hydrazino-3-methylpyrazine
2-Hydrazino-3-methylpyrazine
Vzorec: C₅H₈N₄
MW: 124.144 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19201409
CAS číslo: 19848-54-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR