Ethyl (E)-3-ethoxy-2-methylacrylate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Ethyl (2E)-3-ethoxy-2-methylprop-2-enoate, (E)-Ethyl 3-ethoxy-2-methylacrylate, Ethyl trans-3-ethoxy-2-methylacrylate
OR60192-1G
OR60192-25G
OR60192-5G
APOSOR60192-1GEA
622
CZK
APOSOR60192-1G
APOSOR60192-25G
APOSOR60192-5G
Ethyl (E)-3-ethoxy-2-methylacrylate
Ethyl (E)-3-ethoxy-2-methylacrylate
Vzorec:
C₈H₁₄O₃ MW: 158,195 g/mol Bod varu: 196…198 °C Hustota: 1 g/cm³ (25 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD06204558 CAS číslo: 1085699-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...