Tisk…

8-Bromo-5-chloroquinoline

Dodavatel: Apollo Scientific
Varování

Synonyms: 8-Bromo-5-chloro-1-azanaphthalene

OR60189-1G
APOSOR60189-1GEA 2900 CZK
APOSOR60189-1G
8-Bromo-5-chloroquinoline
8-Bromo-5-chloroquinoline
Vzorec: C₉H₅BrClN
MW: 242,5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193679
CAS číslo: 1154741-20-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR