Tisk…

1-(3-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid

Dodavatel: Apollo Scientific

Synonyms: 4-Carboxy-1-(3-methoxyphenyl)pyrrolidin-2-one

OR60131-1G OR60131-250MG OR60131-500MG OR60131-2.5G OR60131-5G
APOSOR60131-1GEA 2610 CZK
APOSOR60131-1G APOSOR60131-250MG APOSOR60131-500MG APOSOR60131-2.5G APOSOR60131-5G
1-(3-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
Vzorec: C₁₂H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD01630406
CAS číslo: 63674-47-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR