Tisk…

3-Iodo-6-methyl-1H-indazole

Dodavatel: Apollo Scientific
Varování

OR60114-1G OR60114-5G OR60114-250MG
APOSOR60114-1GEA 1740 CZK
APOSOR60114-1G APOSOR60114-5G APOSOR60114-250MG
3-Iodo-6-methyl-1H-indazole
3-Iodo-6-methyl-1H-indazole
Vzorec: C₈H₇IN₂
MW: 258.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781627
CAS číslo: 885518-96-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR